Inositol hexaphosphoric acid dipotassium salt 98%

Supplier: Ambeed
Warning

Synonyms: rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexayl hexakis(dihydrogen phosphate), dipotassium salt, Phytic acid dipotassium salt, Phytic acid potassium

A198387-25G A198387-100G
76800-270EA 20.16 USD
76800-270 77600-612
Inositol hexaphosphoric acid dipotassium salt 98%
Inositol hexaphosphoric acid dipotassium salt

Precaución: for Research Use Only

Formula: C₆H₁₆O₂₄P₆·2K
MW: 736.216 g/mol
MDL Number: MFCD00133773
CAS Number: 129832-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR