Fenticonazole nitrate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-(2-(2,4-Dichlorophenyl)-2-((4-(phenylthio)benzyl)oxy)ethyl)-1H-imidazole nitrate
A198345-100G
A198345-10MG
A198345-1G
A198345-25G
A198345-50MG
A198345-5G
A198345-10G
76800-258EA
361
USD
76800-258
76800-260
76800-262
76800-264
76800-266
76800-268
77879-243
Fenticonazole nitrate 98%
Fenticonazole nitrate
Precaución: for Research Use Only
Formula:
C₂₄H₂₀Cl₂N₂OS·HNO₃ MW: 518.41 g/mol |
MDL Number:
MFCD00941391 CAS Number: 73151-29-8 UN: 2811 ADR: 6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...