(+)-cis-Diltiazem hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (2S,3S)-(+)-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, Diltiazem Hydrochloride, (+)-cis-Diltiazem HCl, (+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, (2S,3S)-5-(2-(Dimethylamino)ethyl)-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl acetate hydrochloride

A198057-1G A198057-1MG A198057-250MG A198057-25G A198057-5G A198057-100G
76800-246EA 28.65 USD
76800-246 76800-248 76800-250 76800-252 76800-254 77600-610
(+)-cis-Diltiazem hydrochloride 95%
(+)-cis-Diltiazem hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₆N₂O₄S·HCl
MW: 450.98 g/mol
MDL Number: MFCD00069252
CAS Number: 33286-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR