Ethyl oxamate 95%

Supplier: Ambeed
Danger

Synonyms: Oxamic Acid Ethyl Ester, Ethyl 2-amino-2-oxoacetate

A197785-100G A197785-10G A197785-25G A197785-500G A197785-5G
76800-236EA 77.45 USD
76800-236 76800-238 76800-240 76800-242 76800-244
Ethyl oxamate 95%
Ethyl oxamate

Precaución: for Research Use Only

Formula: C₄H₇NO₃
MW: 117.1 g/mol
MDL Number: MFCD00008005
CAS Number: 617-36-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR