Meisoindigo 98%

Supplier: Ambeed
Warning

Synonyms: N-Methylisoindigotin, 3-(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)-1,3-dihydro-1-methyl-2H-indol-2-one, 1-Methyl-[3,3'-biindolinylidene]-2,2'-dione

A196338-100MG A196338-10MG A196338-1MG A196338-25MG A196338-50MG A196338-5MG
76800-180EA 135.8 USD
76800-180 76800-182 76800-184 76800-186 76800-188 76800-190
Meisoindigo 98%
Meisoindigo

Precaución: for Research Use Only

Formula: C₁₇H₁₂N₂O₂
MW: 276.29 g/mol
MDL Number: MFCD05988503
CAS Number: 97207-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR