1-(6-Methylpyrimidin-4-yl)ethanone 98%

Supplier: Ambeed
Warning

A195353-100MG A195353-10G A195353-1G A195353-250MG A195353-5G
76800-162EA 35.01 USD
76800-162 76800-164 76800-166 76800-168 76800-170
1-(6-Methylpyrimidin-4-yl)ethanone 98%
1-(6-Methylpyrimidin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.15 g/mol
MDL Number: MFCD09263819
CAS Number: 67073-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR