FCCP 98%

Supplier: Ambeed
Danger

Synonyms: FCCP, [4-(Trifluoromethoxy)phenyl]carbonohydrazonoyl dicyanide, [4-(Trifluoromethoxy)phenylhydrazono]malononitrile, Carbonyl cyanide 4-(Trifluoromethoxy)phenylhydrazone

A194814-100MG A194814-10MG A194814-1MG A194814-5MG
76800-130EA 54.11 USD
76800-130 76800-132 76800-134 76800-136
FCCP 98%
FCCP

Precaución: for Research Use Only

Formula: C₁₀H₅F₃N₄O
MW: 254.17 g/mol
MDL Number: MFCD00009699
CAS Number: 370-86-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR