4-(3-Hydroxypropyl)-2,6-dimethoxyphenol 95%

Supplier: Ambeed
Warning

Synonyms: Dihydrosinapyl alcohol

A194641-100MG A194641-1G A194641-250MG
76800-122EA 120.95 USD
76800-122 77625-962 77625-964
4-(3-Hydroxypropyl)-2,6-dimethoxyphenol 95%
4-(3-Hydroxypropyl)-2,6-dimethoxyphenol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₄
MW: 212.24 g/mol
MDL Number: MFCD00016572
CAS Number: 20736-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR