tert-Butyl-4-methylenepiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-methylenepiperidine, tert-Butyl 4-Methylenepiperidine-1-carboxylate, 4-Methylenepiperidine-1-carboxylic acid tert-Butyl ester, tert-Butyl 4-methylenepiperidine-1-carboxylate, 1-tert-Butoxycarbonyl-4-methylenepiperidine

A194581-10G A194581-1G A194581-250MG A194581-25G A194581-500G A194581-100G
76800-112EA 26.53 USD
76800-112 76800-114 76800-116 76800-118 76800-120 77567-824
tert-Butyl-4-methylenepiperidine-1-carboxylate 98%
tert-Butyl-4-methylenepiperidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₁H₁₉NO₂
MW: 197.2741 g/mol
MDL Number: MFCD01632522
CAS Number: 159635-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR