(R)-1-Boc-piperidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A194273-100G A194273-1G A194273-250MG A194273-25G A194273-5G A194273-500G A194273-10G
76800-084EA 115.99 USD
76800-084 76800-086 76800-088 76800-090 76800-092 77667-626 77879-233
(R)-1-Boc-piperidine-3-carboxylic acid 95%
(R)-1-Boc-piperidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MDL Number: MFCD02179173
CAS Number: 163438-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR