Ethyl-2-chloro-5-oxazolecarboxylate 95%

Supplier: Ambeed
Warning

A193729-100MG A193729-250MG A193729-5G A193729-10G
76800-070EA 14 USD
76800-070 76800-072 76800-074 77567-804
Ethyl-2-chloro-5-oxazolecarboxylate 95%
Ethyl 2-chloro-5-oxazolecarboxylate

Precaución: For research use only.

Formula: C₆H₆ClNO₃
MDL Number: MFCD09475866
CAS Number: 862599-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR