5-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%

Supplier: Ambeed
Warning

A193637-250MG A193637-5G A193637-1G
76800-066EA 5 USD
76800-066 76800-068 77667-610
5-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%
5-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁BrClN
MDL Number: MFCD05861550
CAS Number: 923591-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR