Lithium trifluoromethanesulfonate 95%

Supplier: Ambeed
Warning

A192999-100G A192999-25G A192999-500G A192999-5G
76800-050EA 60 USD
76800-050 76800-052 76800-054 76800-056
Lithium trifluoromethanesulfonate 95%
Lithium trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: CF₃LiO₃S
MW: 156.01 g/mol
MDL Number: MFCD00013227
CAS Number: 33454-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR