Imidazo[1,2-a]pyridine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A192701-100G A192701-1G A192701-25G A192701-5G
76800-036EA 405 USD
76800-036 76800-038 76800-040 76800-042
Imidazo[1,2-a]pyridine-3-carboxylic acid 98%
Imidazo[1,2-a]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD01720458
CAS Number: 6200-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR