Mal-PEG3-acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(2-(2-(2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy)ethoxy)ethoxy)propanoic acid, Maleimido-tri(ethylene glycol)-propionic acid

A192693-50MG A192693-1G A192693-100MG A192693-250MG
76800-034EA 39.26 USD
76800-034 77567-794 77593-238 77667-606
Mal-PEG3-acid 95%
Mal-PEG3-acid

Precaución: For research use only.

Formula: C₁₃H₁₉NO₇
MW: 301.292 g/mol
MDL Number: MFCD23103943
CAS Number: 518044-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR