Diethylamine hydrobromide 98%

Supplier: Ambeed
Warning

A192271-100G A192271-25G A192271-500G
76800-016EA 18.04 USD
76800-016 76800-018 76800-020
Diethylamine hydrobromide 98%
Diethylamine hydrobromide

Precaución: for Research Use Only

Formula: C₄H₁₂BrN
MW: 154.05 g/mol
MDL Number: MFCD00012498
CAS Number: 6274-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR