4'-Bromovalerophenone 97%

Supplier: Ambeed
Warning

A192043-1000G A192043-100G A192043-500G A192043-25G
76800-010EA 129.43 USD
76800-010 76800-012 76800-014 77084-886
4'-Bromovalerophenone 97%
4'-Bromovalerophenone

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO
MW: 241.13 g/mol
MDL Number: MFCD00000107
CAS Number: 7295-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR