Fmoc-N-ME-Trp(boc)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-N-Me-Trp(Boc)-OH

A191514-100MG A191514-1G A191514-250MG A191514-5G
76799-994EA 15 USD
76799-994 76799-996 76799-998 77550-226
Fmoc-N-ME-Trp(boc)-OH 97%
Fmoc-N-ME-Trp(boc)-OH

Precaución: for Research Use Only

Formula: C₃₂H₃₂N₂O₆
MW: 540.616 g/mol
MDL Number: MFCD02094403
CAS Number: 197632-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR