Imidocarb Dipropionate 95%

Supplier: Ambeed
Warning

Synonyms: 1,3-Bis(3-(4,5-dihydro-1H-imidazol-2-yl)phenyl)urea dipropionate

A191373-10G A191373-1G A191373-5G A191373-25G
76799-982EA 57.29 USD
76799-982 76799-984 76799-986 77667-596
Imidocarb Dipropionate 95%
Imidocarb Dipropionate

Precaución: for Research Use Only

Formula: C₁₉H₂₀N₆O·₂C₃H₆O₂
MW: 496.57 g/mol
MDL Number: MFCD00013161
CAS Number: 55750-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR