1-Methyl-1H-indole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A191169-10G A191169-1G A191169-250MG A191169-25G A191169-5G
76799-972EA 26 USD
76799-972 76799-974 76799-976 76799-978 76799-980
1-Methyl-1H-indole-2-carboxylic acid 98%
1-Methyl-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD00005801
CAS Number: 16136-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR