Ethyl 2-amino-1H-indole-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A190527-100MG
A190527-1G
A190527-250MG
A190527-5G
76799-964EA
32
USD
76799-964
76799-966
76799-968
76799-970
Ethyl 2-amino-1H-indole-3-carboxylate 97%
Ethyl 2-amino-1H-indole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₂N₂O₂ |
MDL Number:
MFCD01568663 CAS Number: 6433-72-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...