Diethyl 2-(((3-chlorophenyl)amino)methylene)malonate 95%

Supplier: Ambeed
Warning

A189648-100MG A189648-1G A189648-250MG A189648-5G
76799-948EA 15.92 USD
76799-948 76799-950 76799-952 76799-954
Diethyl 2-(((3-chlorophenyl)amino)methylene)malonate 95%
Diethyl 2-(((3-chlorophenyl)amino)methylene)malonate

Precaución: for Research Use Only

Formula: C₁₄H₁₆ClNO₄
MW: 297.73 g/mol
MDL Number: MFCD00760212
CAS Number: 3412-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR