Meropenem trihydrate 97%
Supplier: Ambeed
|
Danger
|
A188476-100MG
A188476-10G
A188476-1G
A188476-1MG
A188476-250MG
A188476-50MG
A188476-25G
A188476-5G
76799-920EA
8
USD
76799-920
76799-922
76799-924
76799-926
76799-928
76799-930
77600-526
77879-212
Meropenem trihydrate 97%
Meropenem trihydrate
Precaución: for Research Use Only
Formula:
C₁₇H₃₁N₃O₈S |
MDL Number:
MFCD08600005 CAS Number: 119478-56-7 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...