1-(Methylsulfinyl)dodecane 97%

Supplier: Ambeed
Warning

A187578-10G A187578-1G A187578-5G A187578-25G
76799-898EA 159 USD
76799-898 76799-900 76799-902 76799-904 77879-208
1-(Methylsulfinyl)dodecane 97%
1-(Methylsulfinyl)dodecane

Precaución: for Research Use Only

Formula: C₁₃H₂₈OS
MDL Number: MFCD02326311
CAS Number: 3079-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR