4'-Acetamido-2'-methylacetophenone 95%

Supplier: Ambeed
Warning

Synonyms: N-(4-Acetyl-3-methylphenyl)acetamide

A187563-1G A187563-250MG A187563-5G
76799-892EA 33.95 USD
76799-892 76799-894 76799-896
4'-Acetamido-2'-methylacetophenone 95%
4'-Acetamido-2'-methylacetophenone

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.23 g/mol
MDL Number: MFCD02258876
CAS Number: 34956-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR