3-(4-nitrophenyl)-L-alanine ethyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Phe(4-NO2)-OEt·HCl, H-Phe(4-NO2)-OEt.HCl, H-4-Nitro-Phe-OEt·HCl

A186286-100G A186286-25G A186286-5G
76799-880EA 94.43 USD
76799-880 76799-882 76799-884
3-(4-nitrophenyl)-L-alanine ethyl ester hydrochloride 98%
3-(4-nitrophenyl)-L-alanine ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₄·HCl
MW: 274.701 g/mol
MDL Number: MFCD08275409
CAS Number: 58816-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR