6-Amidino-2-naphthol Methanesulfonate 95%
Supplier: Ambeed
|
Warning
|
A185741-10G
A185741-1G
A185741-250MG
A185741-25G
A185741-5G
76799-864EA
192.03
USD
76799-864
76799-866
76799-868
76799-870
76799-872
6-Amidino-2-naphthol Methanesulfonate 95%
6-Amidino-2-naphthol methanesulphonate
Precaución: for Research Use Only
Formula:
C₁₁H₁₀N₂O MW: 282.31 g/mol |
MDL Number:
MFCD00143493 CAS Number: 82957-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...