Chromone-3-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-Oxo-4H-chromene-3-carbaldehyde

A185556-100G A185556-10G A185556-1G A185556-25G A185556-5G
76799-854EA 180.36 USD
76799-854 76799-856 76799-858 76799-860 76799-862
Chromone-3-carboxaldehyde 97%
Chromone-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₆O₃
MW: 174.16 g/mol
MDL Number: MFCD00014667
CAS Number: 17422-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR