Dimethyl-trans-cyclohexane-1,4-dicarboxylate 96%

Supplier: Ambeed
Warning

A185390-100G A185390-10G A185390-1G A185390-25G A185390-5G A185390-500G
76799-844EA 55 USD
76799-844 76799-846 76799-848 76799-850 76799-852 77879-195
Dimethyl-trans-cyclohexane-1,4-dicarboxylate 96%
Dimethyl trans-cyclohexane-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄
MDL Number: MFCD00063917
CAS Number: 3399-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR