Dimethoxymethylphenylsilane 98%

Supplier: Ambeed
Warning

Synonyms: Methylphenyldimethoxysilane, Dimethoxy(methyl)(phenyl)silane

A184573-10G A184573-25G A184573-5G A184573-1000G A184573-500G
76799-830EA 8.49 USD
76799-830 76799-832 76799-834 77625-874 77625-876
Dimethoxymethylphenylsilane 98%
Dimethoxymethylphenylsilane

Precaución: for Research Use Only

Formula: C₉H₁₄O₂Si
MW: 182.29 g/mol
MDL Number: MFCD00008343
CAS Number: 3027-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR