Methyl-2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride 97%

Supplier: Ambeed
Warning

A183127-100G A183127-10G A183127-25G A183127-5G
76799-796EA 81.69 USD
76799-796 76799-798 76799-800 76799-802
Methyl-2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride 97%
Methyl-2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₃
MW: 231.676 g/mol
MDL Number: MFCD00065187
CAS Number: 68697-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR