3-(Pyrimidin-5-yl)benzaldehyde 95%

Supplier: Ambeed
Warning

A182535-1G A182535-250MG
76799-782EA 84.88 USD
76799-782 76799-784
3-(Pyrimidin-5-yl)benzaldehyde 95%
3-(Pyrimidin-5-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C11H8N2O
MW: 184.1900 g/mol
MDL Number: MFCD05864833
CAS Number: 640769-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR