Farnesol (mixture of isomers) 95% (mixture of isomers)
Supplier: Ambeed
|
Danger
|
A182361-1G
A182361-25G
A182361-5G
A182361-100G
A182361-10G
76799-774EA
5
USD
76799-774
76799-776
76799-778
77600-474
77600-476
Farnesol (mixture of isomers) 95% (mixture of isomers)
Farnesol (mixture of isomers)
Precaución: for Research Use Only
Formula:
C₁₅H₂₆O |
MDL Number:
MFCD00002918 CAS Number: 4602-84-0 UN: 3082 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...