5-Methylindol-3-ylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Methyl-1H-indol-3-yl)acetic acid

A182320-250MG A182320-5G
76799-770EA 30.77 USD
76799-770 76799-772
5-Methylindol-3-ylacetic acid 98%
5-Methylindol-3-ylacetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
MDL Number: MFCD00022751
CAS Number: 1912-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR