Isosteviol 98%

Supplier: Ambeed
Warning

Synonyms: (4alpha,8beta,13beta)-13-Methyl-16-oxo-17-norkauran-18-oic acid, (4R,4aS,6aR,9S,11aR,11bS)-4,9,11b-Trimethyl-8-oxotetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carboxylic acid

A182110-100MG A182110-10MG A182110-1MG A182110-5MG
76799-762EA 48.81 USD
76799-762 76799-764 76799-766 76799-768
Isosteviol 98%
Isosteviol

Precaución: for Research Use Only

Formula: C₂₀H₃₀O₃
MW: 318.46 g/mol
MDL Number: MFCD28411514
CAS Number: 27975-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR