1,2-Naphthoquinone-4-sulphonic acid sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Folin's reagent, Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulphonate

A181991-100G A181991-1G A181991-25G A181991-5G
76799-754EA 286 USD
76799-754 76799-756 76799-758 76799-760
1,2-Naphthoquinone-4-sulphonic acid sodium salt 98%
1,2-Naphthoquinone-4-sulphonic acid sodium salt

Precaución: for Research Use Only

Formula: C₁₀H₅NaO₅S
MW: 260.20 g/mol
MDL Number: MFCD00001700
CAS Number: 521-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR