1,2:3,5-Di-O-isopropylidene-ɑ-D-xylofuranose 98+%

Supplier: Ambeed
Warning

Synonyms: (3aR,3bS,7aR,8aR)-2,2,5,5-Tetramethyltetrahydro-7H-[1,3]dioxolo[4',5':4,5]furo[3,2-d][1,3]dioxine

A181940-100G A181940-1G A181940-25G A181940-5G
76799-742EA 320 USD
76799-742 76799-744 76799-746 76799-748
1,2:3,5-Di-O-isopropylidene-ɑ-D-xylofuranose 98+%
1,2:3,5-Di-O-isopropylidene-alpha-D-xylofuranose

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₅
MW: 230.26 g/mol
MDL Number: MFCD00063224
CAS Number: 20881-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR