4-Hydroxy-6-methoxyquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A181855-1G A181855-5G A181855-250MG
76799-738EA 35 USD
76799-738 76799-740 77667-510
4-Hydroxy-6-methoxyquinoline-3-carboxylic acid 97%
4-Hydroxy-6-methoxyquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₄
MDL Number: MFCD00024003
CAS Number: 28027-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR