Methyl-2-bromo-4-methylbenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-bromo-4-methylbenzoate
A181431-100G
A181431-10G
A181431-1G
A181431-25G
A181431-5G
76799-728EA
328.88
USD
76799-728
76799-730
76799-732
76799-734
76799-736
Methyl-2-bromo-4-methylbenzoate 98%
Methyl-2-bromo-4-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol |
MDL Number:
MFCD03840737 CAS Number: 87808-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...