GW-9662 98%

Supplier: Ambeed
Warning

Synonyms: GW9662, 2-Chloro-5-nitro-N-phenylbenzamide

A180779-10MG A180779-1G A180779-250MG A180779-25MG A180779-50MG A180779-5G A180779-5MG A180779-1MG A180779-25G
76799-714EA 8 USD
76799-714 76799-716 76799-718 76799-720 76799-722 76799-724 76799-726 77667-496 77667-498
GW-9662 98%
GW-9662

Precaución: for Research Use Only

Formula: C₁₃H₉N₂O₃Cl
MDL Number: MFCD01215270
CAS Number: 22978-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR