Glycylsarcosine 98%

Supplier: Ambeed
Warning

Synonyms: H-Gly-Sar-OH

A180389-1G A180389-250MG
76799-692EA 54.11 USD
76799-692 76799-694
Glycylsarcosine 98%
Glycylsarcosine

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
MDL Number: MFCD00038193
CAS Number: 29816-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR