2,2-Dimethyl-1-phenylpropylamine 96%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dimethyl-1-phenylpropan-1-amine

A179280-1G A179280-5G
76799-682EA 48.81 USD
76799-682 76799-684
2,2-Dimethyl-1-phenylpropylamine 96%
2,2-Dimethyl-1-phenylpropylamine

Precaución: for Research Use Only

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
MDL Number: MFCD00267378
CAS Number: 61501-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR