2,5-Dihydroxy-1,4-dioxane 95% mixture of stereoisomers.

Supplier: Ambeed
Warning

A179048-100G A179048-10G A179048-1G A179048-25G A179048-5G
76799-664EA 410 USD
76799-664 76799-666 76799-668 76799-670 76799-672
2,5-Dihydroxy-1,4-dioxane 95% mixture of stereoisomers.
2,5-Dihydroxy-1,4-dioxane

Precaución: for Research Use Only

Formula: C₄H₈O₄
MDL Number: MFCD00012133
CAS Number: 23147-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR