Ethyl-4-amino-1H-imidazole-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-amino-1H-imidazole-5-carboxylate

A178870-100MG A178870-1G A178870-250MG A178870-5G
76799-652EA 66.84 USD
76799-652 76799-654 76799-656 76799-658
Ethyl-4-amino-1H-imidazole-5-carboxylate 97%
Ethyl-4-amino-1H-imidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
MDL Number: MFCD06858453
CAS Number: 21190-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR