Ethyl-2-iodobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-iodobenzoate, 2-Iodobenzoic acid ethyl ester

A178861-100G A178861-10G A178861-25G A178861-500G A178861-5G
76799-642EA 94 USD
76799-642 76799-644 76799-646 76799-648 76799-650
Ethyl-2-iodobenzoate 98%
Ethyl-2-iodobenzoate

Precaución: for Research Use Only

Formula: C₉H₉IO₂
MDL Number: MFCD00051796
CAS Number: 1829-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR