Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate 98%

Supplier: Ambeed
Warning

A178627-1000G A178627-100G A178627-500G A178627-25G
76799-632EA 240 USD
76799-632 76799-634 76799-636 77550-150
Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate 98%
Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₃₁H₅₄O₃
MDL Number: MFCD00180757
CAS Number: 67845-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR