3-Butynyl toluene-4-sulfonate 98%

Supplier: Ambeed
Warning

Synonyms: 3-Butynyl 4-methylbenzenesulphonate, 3-Butynyl p-toluenesulphonate

A178595-100G A178595-1G A178595-25G A178595-5G
76799-616EA 550.61 USD
76799-616 76799-618 76799-620 76799-622
3-Butynyl toluene-4-sulfonate 98%
3-Butynyl toluene-4-sulphonate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃S
MW: 224.28 g/mol
MDL Number: MFCD00041687
CAS Number: 23418-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR