3,5-Diiodo-4-methoxybenzonitrile 97%

Supplier: Ambeed
Warning

A178003-1G A178003-250MG
76799-592EA 73 USD
76799-592 76799-594
3,5-Diiodo-4-methoxybenzonitrile 97%
3,5-Diiodo-4-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅I₂NO
MW: 384.94 g/mol
MDL Number: MFCD00144497
CAS Number: 3336-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR