Hexaethyldisiloxane 97%

Supplier: Ambeed
Warning

A177988-100G A177988-10G A177988-1G A177988-25G A177988-5G
76799-582EA 161 USD
76799-582 76799-584 76799-586 76799-588 76799-590
Hexaethyldisiloxane 97%
Hexaethyldisiloxane

Precaución: for Research Use Only

Formula: C₁₂H₃₀OSi₂
MDL Number: MFCD00026693
CAS Number: 994-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR