1-(p-Toluenesulfonyl)pyrrole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Tosylpyrrole, 1-Tosyl-1H-pyrrole, N-Tosylpyrrole

A175752-100G A175752-1G A175752-5G A175752-10G A175752-25G
76799-530EA 163 USD
76799-530 76799-532 76799-534 77879-155 77879-156
1-(p-Toluenesulfonyl)pyrrole 98%
1-(p-Toluenesulfonyl)pyrrole

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂S
MDL Number: MFCD00145014
CAS Number: 17639-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR